Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Seasonal H1N1 Neuraminidase Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344099 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344099 Supplier Novus Biologicals Supplier No. NBP241111MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seasonal H1N1 Neuraminidase Polyclonal antibody specifically detects Seasonal H1N1 Neuraminidase in Influenza virusN1, Virus samples. It is validated for ELISA

Specifications

Antigen Seasonal H1N1 Neuraminidase
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC=3.2.1.18, Neuraminidase
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) Neuraminidase protein. This antibody is a cognate pair with product NBP2-41112. The immunogen is located within the first 50 amino acids of Seasonal H1N1 Neuraminidase. Amino Acid Squence: VAGKDKTSVT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.