Alfa Aesar™ sec-Butyl methyl ether, 99%

Manufacturer: Alfa Aesar™ H53427AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH53427AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6795-87-5
    Molecular Formula C5H12O
    Formula Weight 88.15
    MDL Number MFCD00190205
    UN Number UN2350
    Synonym 2-Methoxybutane
    Chemical Name or Material sec-Butyl methyl ether

    Specifications

    Formula Weight 88.15
    Molecular Formula C5H12O
    MDL Number MFCD00190205
    CAS 6795-87-5
    UN Number UN2350
    Synonym 2-Methoxybutane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 57°C
    Quantity 5mL
    Flash Point −30°C (−22°F)
    RTECS Number KN5249500
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER
    Chemical Name or Material sec-Butyl methyl ether
    Assay 99%

    Description

    Applications
    sec-Butyl methyl ether is used as a solvent in organic synthesis.

    Solubility
    Miscible with water, alcohol, ether and acetone.

    Notes
    Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit