Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC11C Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP179339

 View more versions of this product

Catalog No. NBP179339


Only null left
Add to Cart

Description

Description

SEC11C Polyclonal specifically detects SEC11C in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

SEC11C
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
EC 3.4, Microsomal signal peptidase 21 kDa subunit, SEC11 homolog C, SEC11 homolog C (S. cerevisiae), SEC11L3, SEC11-like protein 3, signal peptidase complex catalytic subunit SEC11C, SPase 21 kDa subunit, SPC21SEC11-like 3 (S. cerevisiae), SPCS4CSEC11-like 3
Rabbit
Affinity purified
RUO
90701
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_079744
SEC11C
The immunogen for this antibody is Sec11c. Peptide sequence VHRVIKVHEKDNGDIKFLTKGDNNEVDDRGLYKEGQNWLEKKDVVGRARG.
100 μL
Primary
Expected identity based on immunogen sequence: Bovine: 100%; Goat: 100%; Human: 100%; Rat: 100%; Xenopus: 92%; Chicken: 78%.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.