Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC11C Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB044998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044998 Supplier Novus Biologicals Supplier No. NBP297566H

Rabbit Polyclonal Antibody

SEC11C Polyclonal specifically detects SEC11C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SEC11C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 3.4, Microsomal signal peptidase 21 kDa subunit, SEC11 homolog C, SEC11 homolog C (S. cerevisiae), SEC11L3, SEC11-like protein 3, signal peptidase complex catalytic subunit SEC11C, SPase 21 kDa subunit, SPC21SEC11-like 3 (S. cerevisiae), SPCS4CSEC11-like 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SEC11C fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90701
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.