Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC13 Antibody (1280B), Alexa Fluor™ 350, R&D Systems™
SDP

Catalog No. MB90551A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB90551A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB90551A350 Supplier Novus Biologicals Supplier No. MAB90551AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SEC13 Monoclonal specifically detects SEC13 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SEC13
Applications Western Blot
Classification Monoclonal
Clone 1280B
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias D3S1231ESEC13L1npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae)
Gene Symbols SEC13
Host Species Rabbit
Immunogen rhSEC13 aa123-322
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6396.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.