Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC22B Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB017948 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017948 0.1 mg
NB10098766 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017948 Supplier Novus Biologicals Supplier No. NB100987660.1MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

SEC22B Polyclonal specifically detects SEC22B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen SEC22B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml
Formulation PBS with No Preservative
Gene Accession No. O75396
Gene Alias ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b
Gene Symbols SEC22B
Host Species Sheep
Immunogen A synthetic peptide as a part of human, rat, mouse, hamster, orangutan, chicken Vesicle-trafficking protein SEC22b conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9554
Test Specificity SEC22B.
Reconstitution Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability.
Target Species Rat, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.