Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC23A Antibody, Novus Biologicals™
SDP

Catalog No. NBP234842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234842 Supplier Novus Biologicals Supplier No. NBP234842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC23A Polyclonal specifically detects SEC23A in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen SEC23A
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry 3 - 5 ug 10^6 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:500
Formulation PBS and 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. B5DFC3
Gene Alias CLSD, MGC26267, protein transport protein Sec23A, Sec23 (S. cerevisiae) homolog A, Sec23 homolog A (S. cerevisiae), SEC23-related protein A
Gene Symbols SEC23A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) T T Y L E F I Q Q N E E R D G V R (18) C of rat COPII.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10484
Test Specificity Cross-reacts with Human, Rat and Mouse. Reported reactivity in Drosopholia.
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.