Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB362790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362790 Supplier Novus Biologicals Supplier No. NBP178752JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEC3 Polyclonal antibody specifically detects SEC3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SEC3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias BM-102, exocyst complex component 1, Exocyst complex component Sec3, FLJ10893, SEC3L1, SEC3-like 1, SEC3-like 1 (S. cerevisiae), SEC3P, SEC3Sec3p
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 100 and 150 of human Exocyst Complex Component Sec3 using the numbering given in entry NP_060731.2 (GeneID 55763).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55763
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.