Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Secretagogin Antibody (048), PerCP, Novus Biologicals™
SDP

Catalog No. NB316884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316884 Supplier Novus Biologicals Supplier No. NBP290100PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Secretagogin Monoclonal antibody specifically detects Secretagogin in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Secretagogin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 048
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CALBL, DJ501N12.8, secretagogin, EF-hand calcium binding protein, SECRETsecretagogin, SEGN, setagin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Secretagogin (Accession#: NP_008929.2; Asp2-Pro276).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10590
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.