Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Secretory phospholipase A2 Type V Antibody, Novus Biologicals™
SDP

Catalog No. NBP231558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231558 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231558 Supplier Novus Biologicals Supplier No. NBP231558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Secretory phospholipase A2 Type V Polyclonal specifically detects Secretory phospholipase A2 Type V in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Secretory phospholipase A2 Type V
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P39877
Gene Alias Ca2+-dependent phospholipase A2, calcium-dependent phospholipase A2, DKFZp686C2294, EC 3.1.1.4, Group V phospholipase A2, GV-PLA2, hVPLA(2), MGC46205, Phosphatidylcholine 2-acylhydrolase 5, phospholipase A2, group V, PLA2-10
Gene Symbols PLA2G5
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GLLDLKSMIEKVTGKNALTNYGFYGCYC
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5322
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.