Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SECTM1 Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB142590 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB142590 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB142590 Supplier Novus Biologicals Supplier No. NBP305817R

Rabbit Polyclonal Antibody

SECTM1 Polyclonal specifically detects SECTM1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SECTM1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human SECTM1 (NP_001618.2; Met 1-Ala 526).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 6398
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.