Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific SecureSeal™ Thermal Adhesive Sealing Film for PCR Application

Catalog No. 23045247 Shop All Simport Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
23-045-247 Non-sterile
23-045-248 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-045-247 Supplier Simport™ Scientific Supplier No. T3291
Add to Cart
Add to Cart

SecureSeal™ Thermal Adhesive Sealing Film for PCR Application consists of 2.0 mil polyolefin film coated on one side with pressure sensitive acrylate adhesive, which does not interfere with cycle reactions.

  • Low-auto fluorescence
  • Thermostable and functional from -70°C to +100°C
  • Certified RNAse, DNAse and DNA-free
  • DMSO resistant

Specifications

Color Transparent
Certifications/Compliance RNase, Dnase and DNA-free
Material Polyolefin
Sterility Non-sterile
For Use With (Application) PCR
Product Type Sealing Film
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.