Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELENBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155263 Supplier Novus Biologicals Supplier No. NBP155263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SELENBP1 Polyclonal specifically detects SELENBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SELENBP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q13228
Gene Alias 56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813
Gene Symbols SELENBP1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SELENBP1(selenium binding protein 1) The peptide sequence was selected from the C terminal of SELENBP1 (AAH32997). Peptide sequence KQFYPDLIREGSVMLQVDVDTVKGGLKLNPNFLVDFGKEPLGPALAHELR.
Molecular Weight of Antigen 44 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8991
Test Specificity Recognizes C-terminal region on both isoforms of SELENBP1.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.