Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELENBP1 Antibody (OTI3H1), FITC, Novus Biologicals™
SDP

Catalog No. NBP274053F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274053F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274053F Supplier Novus Biologicals Supplier No. NBP274053F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SELENBP1 Monoclonal specifically detects SELENBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen SELENBP1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3H1
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813
Gene Symbols SELENBP1
Host Species Mouse
Immunogen Full length recombinant protein of human SELENBP1 (NP_003935) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8991.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.