Learn More
Selenourea, 99%
Description
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.
Solubility
Soluble in water, alcohol and ether.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽210°C (decomposition) |
| Assay Percent Range | 99+% metals basis |
| Quantity | 1 g |
| UN Number | UN3283 |
| Sensitivity | Air, light, and moisture sensitive |
| Solubility Information | Soluble in water,alcohol and ether. |
| Formula Weight | 123.02 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | Selenourea |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Hazard Statement | GHS H Statement H301-H331-H373 |
| Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
| EINECSNumber | 211-129-9 |
| RTECSNumber | YU1820000 |
| TSCA | Yes |
| storageNote1 | Air and Moisture Sensitive |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.