Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Selinexor
GREENER_CHOICE

Catalog No. p-7249492
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective exportin 1 (XPO1/CRM1) inhibitor

Selinexor is an inhibitor of the exportin 1 (XPO1/CRM1) nuclear exporter. Shows selectivity for TNBC over ER-positive breast cancer cell lines (IC50 values are 11-550 nM and >1000 nM, respectively), and reduces growth of xenografts. Inhibits growth of thyroid cancer cell lines (IC50 = 150-500 nM), potentiates the anti-tumor effects of Olaparib (Cat. No. 7579) in ovarian cancer and breast cancer. Also a potent inhibitor of SARS-CoV-2 replication in vitro (EC50 = 10 nM) and downregulates the production of pro-inflammatory cytokines including TNF-α and IL-6.

Specifications

Chemical Name or Material (2Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyrazinyl)hydrazide
CAS 1393477-72-9
Quantity 10 mg
Target Antiviral Compounds
Molecular Formula C17H11F6N7O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.