Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SELK Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB293087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293087 Supplier Novus Biologicals Supplier No. NBP326091AF488

Rabbit Polyclonal Antibody

SELK Polyclonal antibody specifically detects SELK in Human samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen SELK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias HSPC297, MGC17057, selenoprotein K, SelK
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SELK fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58515
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.