Description
- Clear UV-absorbing polycarbonate lens
- Soft vinyl goggle frame conforms to facial contours for a secure fit
- Protect from flying particles
- Fit comfortably over most prescription eyewear
- Direct venting ensures airflow and cools the eye area
Note:Do not use these goggles for splash protection.
Order Info
Available in standard or fog-free version.
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Impact Goggles |
| Material | Polycarbonate |
| Frame Material | Vinyl |