Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Selpercatinib
GREENER_CHOICE

Catalog No. p-7250117
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective RET inhibitor

Selpercatinib, is a potent and selective receptor tyrosine kinase RET inhibitor (EC50 = 4 nM). It shows > 100-fold selectivity for > 95% of 228 purified kinase domains in radiometric assays. In vitro, it inhibits proliferation of cancer cell lines containing RET mutations or gene fusions. In vivo, it inhibits tumor growth in diverse RET mouse models. Selpercatinib is orally bioavailable.

Specifications

Chemical Name or Material 6-(2-Hydroxy-2-methylpropoxy)-4-[6-[6-[(6-methoxy-3-pyridinyl)methyl]-3,6-diazabicyclo[3.1.1 ]hept-3-yl]-3-pyridinyl]pyrazolo[1,5-a]pyridine-3-carbonitrile
CAS 2152628-33-4
Quantity 10 mg
Target Ret
Molecular Formula C29H31N7O3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.