Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA5A Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $481.00
Specifications
Antigen | SEMA5A |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551131
|
Invitrogen™
MA551131 |
200 μL |
Each for $481.00
|
|
|||||
FEMA551132
|
Invitrogen™
MA551132 |
20 μL |
Each for $195.80
|
|
|||||
Description
Members of the semaphorin protein family, such as SEMA5A, are involved in axonal guidance during neural development (Adams et al., 1996 [PubMed 8817451]).Specifications
SEMA5A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
5930434A13; 9130201M22Rik; AI464145; Ensembl:ENSRNOP00000016506}; M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; sema F; SEMA5A; sema5a {ECO:0000312; SEMAF; semaphorin 5A; semaphorin F; semaphorin-5A; semaphorin-5A {ECO:0000250; Semaphorin-F; semF; UniProtKB:Q13591} | |
SEMA5A | |
IgG1 κ | |
Protein A/G | |
SEMA5A |
C3 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
D3ZTD8, Q13591 | |
310207, 9037 | |
Recombinant Human SEMA5A protein, Val319-Gln668 (Accession #Q13591). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title