Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3A Antibody (1040525), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB297524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297524 Supplier Novus Biologicals Supplier No. FAB109031Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Semaphorin 3A Monoclonal specifically detects Semaphorin 3A in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Semaphorin 3A
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1040525
Conjugate DyLight 755
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias (semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD
Host Species Rat
Immunogen Chinese Hamster Ovary cell line CHO-derived mouse Semaphorin 3A protein, Asn21-Lys747, Accession # O08665
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10371
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.