Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3A Antibody, Novus Biologicals™
SDP

Catalog No. NB387854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387854 Supplier Novus Biologicals Supplier No. NBP323379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Semaphorin 3A Polyclonal antibody specifically detects Semaphorin 3A in Human samples. It is validated for Western Blot

Specifications

Antigen Semaphorin 3A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias (semaphorin) 3A, coll-1, collapsin 1, Hsema-I, Hsema-III, MGC133243, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema III, SEMA1, SEMAD, SEMAIII, SEMAL, semaphorin D, Semaphorin III, semaphorin L, semaphorin-3A, SemD
Host Species Rabbit
Immunogen Semaphorin 3A synthetic peptide (coupled to carrier protein) corresponding to amino acids in the C-terminal region of human Semaphorin 3A. The sequence used has two amino acid differences from rat and mouse Semaphorin 3A, and has low homology to other semaphorin family members. [UniProt# Q14563]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10371
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.