Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3B Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB1002218H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002218H 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002218H Supplier Novus Biologicals Supplier No. NB1002218H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Semaphorin 3B Polyclonal specifically detects Semaphorin 3B in Bovine, Canine, Human, Mouse, Primate, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Semaphorin 3B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Accession No. Q13214
Gene Alias (semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, semaV
Gene Symbols SEMA3B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200). [UniProt# Q13214]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7869
Test Specificity Mouse, human, rat, dog, bovine, and primate.
Target Species Bovine, Canine, Human, Mouse, Primate, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.