Learn More
Semaphorin 3B Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 2 publications
Supplier:  Novus Biologicals NB1002218SS
 View more versions of this product
Catalog No. NB1002218SS
Only null left
Add to Cart
Description
Description
Semaphorin 3B Polyclonal specifically detects Semaphorin 3B in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Semaphorin 3B
Polyclonal
Unconjugated
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
(semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, semaV
Rabbit
Affinity Purified
RUO
Primary
Mouse, human, rat, dog, bovine, and primate.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:5000-1:20000, Simple Western 1:25, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin 1:100 - 1:200
Q13214
SEMA3B
A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200). [UniProt# Q13214]
0.025 mL
Cancer, Growth and Development, Neuronal Cell Markers
7869
Human, Mouse, Rat, Bovine, Canine, Primate
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only