Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Semaphorin 3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$268.00 - $512.00
Specifications
Antigen | Semaphorin 3B |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:5000-1:20000, Simple Western 1:25, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin 1:100 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002218SS
![]() |
Novus Biologicals
NB1002218SS |
0.025 mL |
Each for $268.00
|
|
|||||
NB1002218
![]() |
Novus Biologicals
NB1002218 |
0.05 mL |
Each for $512.00
|
|
|||||
Description
Semaphorin 3B Polyclonal specifically detects Semaphorin 3B in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Semaphorin 3B | |
Western Blot 1:5000-1:20000, Simple Western 1:25, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Growth and Development, Neuronal Cell Markers | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
(semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, semaV | |
SEMA3B | |
IgG | |
Affinity Purified | |
Mouse, human, rat, dog, bovine, and primate. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine, Canine, Primate | |
Q13214 | |
7869 | |
A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200). [UniProt# Q13214] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title