Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3C Antibody (RM0315-7F18), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP211877A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP211877A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP211877A94 Supplier Novus Biologicals Supplier No. NBP211877AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Semaphorin 3C Monoclonal specifically detects Semaphorin 3C in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Semaphorin 3C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0315-7F18
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias (semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE
Gene Symbols SEMA3C
Host Species Rat
Immunogen Mouse recombinant Semaphorin 3C
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10512
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.