Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3C Antibody (RM0315-7F18), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361615 Supplier Novus Biologicals Supplier No. NBP211877MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Semaphorin 3C Monoclonal antibody specifically detects Semaphorin 3C in Mouse samples. It is validated for Western Blot

Specifications

Antigen Semaphorin 3C
Applications Western Blot
Classification Monoclonal
Clone RM0315-7F18
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias (semaphorin) 3C, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema E, SEMAE, semaphorin E, semaphorin-3C, Semaphorin-E, SemE
Host Species Rat
Immunogen Mouse recombinant Semaphorin 3C
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10512
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.