Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 3E Antibody (400513), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB304070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304070 Supplier Novus Biologicals Supplier No. FAB32391N

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Semaphorin 3E Monoclonal antibody specifically detects Semaphorin 3E in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Semaphorin 3E
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 400513
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, SEMAHM-SemaK, semaphorin-3E
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E, Thr25-Ser775 (Arg557Ala and Arg560Ala), Accession # O15041
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9723
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.