Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4B Antibody (561416), PerCP, Novus Biologicals™
SDP

Catalog No. NB367936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367936 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367936 Supplier Novus Biologicals Supplier No. FAB5485C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorin 4B Monoclonal specifically detects Semaphorin 4B in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Semaphorin 4B
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 561416
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias KIAA1745sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4B, MGC131831, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4B, SEMAC, semaphorin-4B, SemC
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4B, Leu39-Glu712, Accession # Q9NPR2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10509
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.