Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4D/CD100 Antibody (010), DyLight 488
SDP

Catalog No. NB100520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100520 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100520 Supplier Novus Biologicals Supplier No. NBP290666G

Rabbit Monoclonal Antibody

Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Semaphorin 4D/CD100
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 10
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Semaphorin 4D/CD100 (NP_038688.2; Met1-Arg733).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.