Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4D/CD100 Antibody (133-1C6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247795B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247795B 0.2 mg
NBP247795A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247795B Supplier Novus Biologicals Supplier No. NBP2477950.2MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Semaphorin 4D/CD100 Monoclonal antibody specifically detects Antigen in Baboon, Equine (Negative), Human samples. It is validated for Flow Cytometry, Immunofluorescence.

Specifications

Antigen Semaphorin 4D/CD100
Applications Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 133-1C6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Gene Symbols SEMA4D
Host Species Mouse
Immunogen PHA stimulated human peripheral blood lymphocytes.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10507
Test Specificity Recognizes a homodimeric protein comprised of 50kDa subunits, identified as CD100 (Workshop VI; Code N-L026). It is expressed on majority of haemopoietic cells (B, T, NK and myeloid cells) and is absent from bone marrow, erythrocytes, eosinophils and endothelial cells. Its expression is increased after PHA-activation. CD100 was shown to associate with different partner molecules in T cells such as CD45, a key molecule with protein tyrosine phosphatase activity involved in T-cell transduction, and a Serine kinase. It plays a role in homotypic cell adhesion and in T cell activation.
Target Species Human, Baboon, Equine (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.