Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4D/CD100 Antibody (758726), PerCP, Novus Biologicals™
SDP

Catalog No. NB374605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374605 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374605 Supplier Novus Biologicals Supplier No. FAB74701C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Semaphorin 4D/CD100
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 758726
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Semaphorin 4D/CD100, Met22-Arg734, Accession # Q92854
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.