Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4D/CD100 Antibody (A8), PE, Novus Biologicals™
SDP

Catalog No. NBP225198PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225198PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225198PE Supplier Novus Biologicals Supplier No. NBP225198PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen Semaphorin 4D/CD100
Applications Flow Cytometry
Classification Monoclonal
Clone A8
Conjugate PE
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation PBS with 0.05% Sodium Azide
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Gene Symbols SEMA4D
Host Species Mouse
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.