Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4D/CD100 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345003 Supplier Novus Biologicals Supplier No. NB10093562MFV450

Rabbit Polyclonal Antibody

Semaphorin 4D/CD100 Polyclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Western Blot

Specifications

Antigen Semaphorin 4D/CD100
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 777-788 [KPKSDFCDREQS] of the human CD100 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.