Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 4F Antibody (780225), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB375003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375003 Supplier Novus Biologicals Supplier No. FAB7200D
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Semaphorin 4F Monoclonal specifically detects Semaphorin 4F in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Semaphorin 4F
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 780225
Conjugate DyLight 350
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias M-SEMA, m-Sema-M, PRO2353, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4F, Sema M, Sema W, SEMAM, semaphorin-4F, semaphorin-M, Semaphorin-W, SEMAWsemaphorin M
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F, Arg41-Gly667, Accession # Q9Z123
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10505
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.