Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 6B Antibody (MM0535-10F7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP211880A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP211880A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP211880A32 Supplier Novus Biologicals Supplier No. NBP211880AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorin 6B Monoclonal specifically detects Semaphorin 6B in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 6B
Applications Western Blot
Classification Monoclonal
Clone MM0535-10F7
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353
Gene Symbols SEMA6B
Host Species Mouse
Immunogen Human recombinant Semaphorin 6B (extracellular domain)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10501
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.