Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 6B Antibody (MM0535-10F7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB361610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361610 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361610 Supplier Novus Biologicals Supplier No. NBP211880APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Semaphorin 6B Monoclonal antibody specifically detects Semaphorin 6B in Human samples. It is validated for Western Blot

Specifications

Antigen Semaphorin 6B
Applications Western Blot
Classification Monoclonal
Clone MM0535-10F7
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353
Host Species Mouse
Immunogen Human recombinant Semaphorin 6B (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10501
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.