Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Semaphorin 7A Antibody (MM0537-99D22), HRP, Novus Biologicals™
SDP

Catalog No. NBP211884H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211884H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211884H Supplier Novus Biologicals Supplier No. NBP211884H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Semaphorin 7A Monoclonal specifically detects Semaphorin 7A in Human samples. It is validated for Western Blot.

Specifications

Antigen Semaphorin 7A
Applications Western Blot
Classification Monoclonal
Clone MM0537-99D22
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, SEMAK1, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L
Gene Symbols SEMA7A
Host Species Rat
Immunogen Human recombinant Semaphorin 7A (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8482
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.