Learn More
Description
Specifications
Specifications
| Antigen | Semenogelin I |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | CT103, dJ172H20.2, FLJ78262, semen coagulating protein, semenogelin IMGC14719, semenogelin-1, SEMGcancer/testis antigen 103, SGI |
| Gene Symbols | SEMG1 |
| Host Species | Rabbit |
| Immunogen | Gel purified rat p47 seminal vesicle protein I (secreted Semenogelin I) from rodent seminal vesicle secretion fluid was used as immunogen. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
