Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Semi-circular Lens 75 mm Base
SDPSCI_ED

Catalog No. S15960 Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Product Type: Lens

Ideal for use with various optics demonstrations.

  • Acrylic semi-circular lens has a 75 mm base and is 15 mm thick.

Specifications

Material Acrylic
Product Type Lens
Shape Semi-circular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.