Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Senataxin Antibody (4G1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252726SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252726SS 0.025 mg
NBP252726 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP252726SS Supplier Novus Biologicals Supplier No. NBP252726SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Senataxin Monoclonal specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Senataxin
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4G1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 5 ug/ml, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500, Flow (Intracellular) 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q7Z333
Gene Alias ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX
Gene Symbols SETX
Host Species Mouse
Immunogen Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt# Q7Z333]
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23064
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.