Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Senataxin Antibody (4G1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Senataxin |
---|---|
Clone | 4G1 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 5 ug/ml, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500, Flow (Intracellular) 5 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP252726SS
![]() |
Novus Biologicals
NBP252726SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP252726
![]() |
Novus Biologicals
NBP252726 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Senataxin Monoclonal specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Senataxin | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
DNA Repair | |
PBS with 0.05% Sodium Azide | |
ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
SETX | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4G1 | |
Flow Cytometry 5 ug/ml, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500, Flow (Intracellular) 5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q7Z333 | |
23064 | |
Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt# Q7Z333] | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title