Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Senataxin Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N10743A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10743A70 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10743A70 Supplier Novus Biologicals Supplier No. NB10057543AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for ChIP Assay, Immunoprecipitation, KnockDown, Western Blot (Negative).

Specifications

Antigen Senataxin
Applications ChIP Assay, Immunoprecipitation, KnockDown, Western Blot (Negative)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Knockout Validated, KnockDown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX
Gene Symbols SETX
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23064
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.