Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP197417SS
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative), Immunofluorescence.Specifications
Vanilloid R1/TRPV1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Invertebrate, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
1.0 mg/ml | |
Western Blot 1:1000, Immunohistochemistry reported by customer review, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry (Negative), Immunofluorescence | |
O35433 | |
TRPV1 | |
A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] | |
0.025 mL | |
Neuroscience, Sensory Systems | |
7442 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction