Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$207.00 - $499.50
Specifications
Antigen | Vanilloid R1/TRPV1 |
---|---|
Concentration | 1.0 mg/ml |
Dilution | Western Blot 1:1000, Immunohistochemistry reported by customer review, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry (Negative), Immunofluorescence |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197417SS
![]() |
Novus Biologicals
NBP197417SS |
0.025 mL |
Each for $207.00
|
|
|||||
NBP197417
![]() |
Novus Biologicals
NBP197417 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Negative), Immunofluorescence.Specifications
Vanilloid R1/TRPV1 | |
Western Blot 1:1000, Immunohistochemistry reported by customer review, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry (Negative), Immunofluorescence | |
Polyclonal | |
Rabbit | |
Neuroscience, Sensory Systems | |
PBS with 0.02% Sodium Azide | |
Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
TRPV1 | |
IgG | |
Affinity Purified |
1.0 mg/ml | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Invertebrate, Primate | |
O35433 | |
7442 | |
A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title