Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Senataxin Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291765 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291765 Supplier Novus Biologicals Supplier No. NB10057543PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Senataxin Polyclonal antibody specifically detects Senataxin in Human samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP), KnockDown, KnockDown

Specifications

Antigen Senataxin
Applications Western Blot, Immunoprecipitation, ChIP Assay, KnockDown, KnockDown
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated
Formulation PBS
Gene Alias ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX
Host Species Rabbit
Immunogen Maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23064
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.