Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056405 0.1 mg
NB10056405T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056405 Supplier Novus Biologicals Supplier No. NB10056405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

SENP1 Polyclonal specifically detects SENP1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen SENP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 - 5 ug/ml, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27882949), Immunohistochemistry-Paraffin reported in scientific literature (PMID 27882949)
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.4.22, EC 3.4.22.-, Sentrin/SUMO-specific protease SENP1, sentrin-specific protease 1, SUMO1/sentrin specific peptidase 1, SUMO1/sentrin specific protease 1, SuPr-2
Gene Symbols SENP1
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to aa 578-590 of human SENP1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29843
Test Specificity A 66 kDa band is observed. In human and mouse, SENP1 has different isoforms between 664 and 699 amino acids long. Additional bands seen in western blot may represent other SUMOlated proteins.
Target Species Human, Mouse, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.