Learn More
Description
Specifications
Specifications
| Antigen | SENP1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 3 - 5 ug/ml, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27882949), Immunohistochemistry-Paraffin reported in scientific literature (PMID 27882949) |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | EC 3.4.22, EC 3.4.22.-, Sentrin/SUMO-specific protease SENP1, sentrin-specific protease 1, SUMO1/sentrin specific peptidase 1, SUMO1/sentrin specific protease 1, SuPr-2 |
| Gene Symbols | SENP1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
