Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP1 Rabbit anti-Human, Clone: 1C3I4, Novus Biologicals™
SDP

Catalog No. NB167926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167926 20 μg
NB167925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167926 Supplier Novus Biologicals Supplier No. NBP31641720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SENP1 Monoclonal antibody specifically detects SENP1 in Human samples. It is validated for Western Blot

Specifications

Antigen SENP1
Applications Western Blot
Classification Monoclonal
Clone 1C3I4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 3.4.22, EC 3.4.22.-, Sentrin/SUMO-specific protease SENP1, sentrin-specific protease 1, SUMO1/sentrin specific peptidase 1, SUMO1/sentrin specific protease 1, SuPr-2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-501 of human SENP1 (Q9P0U3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 29843
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.