Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP2 Antibody (OTI4B3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274060B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274060B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274060B Supplier Novus Biologicals Supplier No. NBP274060B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

SENP2 Monoclonal specifically detects SENP2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SENP2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4B3
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2
Gene Symbols SENP2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids SENP2(NP_067640) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 59343.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.